Learn More
Thermo Scientific™ Agglutinating Sera, Salmonella O
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Brand: Thermo Scientific™ R30957801
Description
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions

Specifications
Positive | |
Salmonella O Antigen | |
Group H | |
2 mL |
Salmonella O Factor 14 (group H) | |
Agglutinating Sera | |
2 mL/Vial | |
Salmonella O |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3212932 | Certificate of Analysis | 30/07/2021 | R30957801 |
3163769 | Certificate of Analysis | 23/11/2020 | R30957801 |
2912566 | Certificate of Analysis | 03/06/2020 | R30957801 |
2843617 | Certificate of Analysis | 14/10/2019 | R30957801 |
2443543 | Certificate of Analysis | 17/01/2019 | R30957801 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.