Learn More
3M™ Reusable Full Face Respirators
An economical and practical choice for filtering out a variety of hazardous materials.
Brand: 3M™ 7000108594
Description
- Used with twin lightweight filters attached by a simple bayonet attachment system
- Flexible system (gas/vapor and/or particulate filters plus Supplied-Air option)
- Soft, pliable, hypo-allergenic facepiece material
- Easy to use
- Dual filter design reduces breathing resistance and improves weight distribution for greater comfort
- Large, polycarbonate lens
- Double face sealing gasket ensures a better fit
- Speech diaphragm improves communication
- Six strap harness assembly for easy adjustment and a reliable fit
- Soft compound inner nose-cup increases user comfort
- Spectacle kit available
Specifications
Bayonet | |
Reusable Full Face Mask Respirator | |
Polycarbonate visor, six strap head harness, cool-flow valve, silicone rubber facepiece | |
1 Ea. |
Black | |
6 strap assembly | |
Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed.