Learn More
Labcon Reusable Racks For Centrifuge Tubes
Plastic centrifuge tube storage racks eliminate wasteful styrofoam racks. They are durable, reusable, interlockable, stackable, freezable and recyclable. Made from 100% pre-consumer, high quality recycled polypropylene from our in-house scrap recycling program.
37.90 € - 59.80 €
Specifications
Product Type | Test Tube Rack |
---|---|
Includes | Racks |
Array | 25 |
Autoclavable | Autoclavable |
Holds | 25 Tubes |
Product Code | Brand | Dimensions (L x W) | For Use With (Equipment) | Color | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | For Use With (Equipment) | Color | Height (Metric) | Price | Quantity & Availability | ||
17494342
|
Labcon
2715-220-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Natural | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17404352
|
Labcon
2715-221-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | White | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17414352
|
Labcon
2715-222-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Yellow | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17424352
|
Labcon
2715-223-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Orange | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17434352
|
Labcon
2715-224-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Red | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17444352
|
Labcon
2715-226-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Purple | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17454352
|
Labcon
2715-227-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Blue | 3.81 cm |
42.15 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17464352
|
Labcon
2715-228-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Green | 3.81 cm |
38.95 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17474352
|
Labcon
2715-229-000-9 |
14.88 x 11.7 cm | 15 mL Centrifuge Tubes and 5 mL Snap Cap Tubes | Assorted | 3.81 cm |
59.80 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17484352
|
Labcon
2750-220-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Natural | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17494352
|
Labcon
2750-221-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | White | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17404362
|
Labcon
2750-222-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Yellow | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17414362
|
Labcon
2750-223-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Orange | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17424362
|
Labcon
2750-224-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Red | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17434362
|
Labcon
2750-226-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Purple | 3.18 cm |
49.25 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17444362
|
Labcon
2750-227-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Blue | 3.18 cm |
45.65 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17454362
|
Labcon
2750-228-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Green | 3.18 cm |
37.90 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
17464362
|
Labcon
2750-229-000-9 |
21.29 x 18.06 cm | 50 mL Centrifuge Tubes | Assorted | 3.18 cm |
59.80 €
Pack of 5 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
Description
Designed to hold Labcon™ tubes specifically, but may fit other brands of centrifuge tubes.
- Holds (25) 15 or 50 mL Conical Centrifuge Tubes
- Includes Numbered Wells
- Racks Lock Together To Form Work Stations
- Replaces Disposable Polystyrene Foam Racks
- Made of Polypropylene
- Freezable to -80°C
- Autoclavable
Specifications
Test Tube Rack | |
25 | |
25 Tubes | |
Yes | |
Freezable (-80°C) |
Racks | |
Autoclavable | |
Polypropylene | |
Non-sterile |