Learn More
Thermo Scientific™ Rheodyne™ Stators
Use these Thermo Scientific; Rheodyne; Stators for your Rheodyne Sample Injectors.
Brand:  Thermo Scientific™ 7010-040
638.32 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10056264
  • 641.51 € / Each
Explore more special offers
Description
Description
Use Thermo Scientific™ Rheodyne™ Stators with popular Rheodyne injector models used in high-performance liquid chromatography (HPLC) systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stators
For Rheodyne injector models 7000/7010/7030/7040/7125
Rheodyne Injector Models 7000/7010/7030/7040/7125
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Rheodyne™ Stators > For use with Rheodyne injector models 7000/7010/7030/7040/7125
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.