missing translation for 'onlineSavingsMsg'
Learn More
Riboflavin, 98%, Thermo Scientific Chemicals Product Code.: 11489323

Riboflavin, 98%, Thermo Scientific Chemicals

Product Code. 11489323
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11489323

Brand: Thermo Scientific Alfa Aesar A11764.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Photoinitiator of polymerization for PAGE gelsRiboflavin is used in the preparation of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). It also provides protection against oxidative damage. It is also used as a food coloring and a vitamin. Further, it is employed as a medication, animal feed supplement and nutritional supplement. In addition to this, it is used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Photoinitiator of polymerization for PAGE gelsRiboflavin is used in the preparation of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). It also provides protection against oxidative damage. It is also used as a food coloring and a vitamin. Further, it is employed as a medication, animal feed supplement and nutritional supplement. In addition to this, it is used as a flavoring agent.

Solubility
Soluble in water, absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform.

Notes
Store in a cool place. Incompatible with Strong oxidizing agents, reducing agents, bases, calcium and metallic salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-88-5
Molecular Formula C17H20N4O6
Molecular Weight (g/mol) 376.37
MDL Number MFCD00005022
InChI Key AUNGANRZJHBGPY-QTZZOOGMNA-N
Synonym riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin
PubChem CID 71310809
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
SMILES CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point 290°C
Assay Percent Range 98%
Odor Odorless
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,8200
Solubility Information Soluble in water,absolute ethanol and dimethyl sulfoxide. Insoluble in acetone and chloroform.
Formula Weight 376.37
Percent Purity 98%
Chemical Name or Material Riboflavin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462723 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberA11764.14,A11764.22
Lot NumberY14J028 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog NumberA11764.14,A11764.22
Lot NumberY26K015 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberA11764.14,A11764.22
Lot NumberW13K025 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberA11764.14,A11764.22
Lot Number10250215 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA11764.14,A11764.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riboflavin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.