Learn More
3M™ Secure Click™ Quantitative Fit Test Adaptor Kit HF-800-03
Kit for conducting quantitative fit testing on 3M™ Secure Click™ Half Mask Reusable Respirators HF-800SD Series. Cartridges and filters are not included.
Brand:  3M™ 7100181558
74.92 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17217680
  • 201.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Use to conduct quantitative fit testing on 3M™ Secure Click™ Half Mask Reusable Respirators with speaking diaphragm
  • Includes head harness assembly
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter Kit
Conducting Quantitative Fit Testing on 3 M™ Secure Click™ Half Mask Reusable Respirators HF-800 SD Series
Head harness assembly
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Secure Click™ Quantitative Fit Test Adaptor Kit HF-800-03 > Head harness assembly