missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
9635.00 € - 19415.00 €
Specifications
Capacity (English) Shelf | 22 lb. |
---|---|
Capacity (Metric) Shelf | 10 kg |
Chamber Material | Stainless Steel |
CO2 Concentration Range | 0 to 20% |
CO2 Sensor Technology | IR Sensor |
Product Code | Brand | Capacity (Metric) | Model | Oxygen Control | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Model | Oxygen Control | Price | Quantity & Availability | |||
17383693
|
BINDER™
9640-0005 |
53 L | CB056-230 V | None |
9635.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17393693
|
BINDER™
9640-0006 |
53 L | CB056-230 V-O | Yes | N/A | ||||
17739794
|
BINDER™
9640-0027 |
170 L | CB170-230V-D | None |
13160.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
17303703
|
BINDER™
9640-0009 |
170 L | CB170-230 V | None |
12590.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17313703
|
BINDER™
9640-0010 |
170 L | CB170-230 V-O | Yes |
14710.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17323703
|
BINDER™
9640-0013 |
267 L | CB260-230 V | None |
17110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17333703
|
BINDER™
9640-0014 |
267 L | CB260-230 V-O | Yes |
19415.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Optimized double-pan humidification system with condensation protection
- Tried-and-tested anti-contamination concept with 180°C hot air sterilization
- Hot-air sterilizable CO2 sensor with single-beam infrared technology
- Seamless inner chamber made of stainless steel with flanges as shelf support system
- Fanless interior with Venturi CO2 gas mixing nozzle
- Intuitive touchscreen controller
- Internal data recording, can be read out in open format via USB interface
- Fault diagnosis system with visual and acoustic alarm
- Interfaces: Ethernet, USB, zero-voltage alarm contact
- Fail-safe CO2 system for protecting the pH of cell cultures
Specifications
22 lb. | |
Stainless Steel | |
IR Sensor | |
None | |
1 | |
90 to 95% | |
TFT touch | |
IR sensor |
10 kg | |
0 to 20% | |
Natural | |
No Lighting | |
Nema 5-20P | |
CO2 Incubator | |
IR sensor | |
50/60 Hz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title