Learn More
SGE™ GC Inlet FocusLiners™
Results in lower mass discrimination
Brand: SGE™ 092002
Description
Results in lower mass discrimination
- For various GC Instruments
- All liners are deactivated using high temperature (420°C) gas phase deactivation
- Supplied in blister packs to prevent contamination
- Reduced active sites mean no unwanted reaction in samples
- Includes two tapered sections that secure quartz wool plug for optimum needle tip wiping and sample transfer, also prevents quartz wool from shifting toward septum or pushing down into liner
- Provide excellent reproducibility
Specifications
5/Pk. | |
Quartz Wool | |
0.4 cm | |
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177 |
7.85 cm | |
Split/Splitless Fast Focus Liner | |
0.63 cm |
Product Suggestions
Customers who viewed this item also viewed.