Learn More
Trajan™ Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Fixed needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves
Brand:  Trajan™ 006300
 View more versions of this product
Product Code. 10209060
  • 74.45 € / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Specifications
Borosilicate Glass (Barrel)
0.37 mm
0.028 mm
1/Ea.
LC
22 G
6.5 mm
Manual Syringe
250 μL
Beckman/Altex, Rheodyne, SSI, Valco Valves
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan™ Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves > for Rheodyne/Valco valves HPLC tip fixed needle 250μL 2 x 0.028 inch needle length x outer diameter SGE