missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 3.5K MWCO

Perform sample buffer exchange with these 3500-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Brand:  Thermo Scientific™ 88403

309.70 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11552571

  • 333.00 € / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Cup, Conical
3.5 kDa
MINI Dialysis Device
Buffer Exchange, Protein
25 Samples
Store at room temperature
Slide-A-Lyzer™
Cellulose Membrane, Dialysis
MINI Dialysis Device
2 mL
25 Devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP00091094 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number88403
Lot NumberP00091056 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number88403
Lot NumberP00091032 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number88403
Lot NumberP00091025 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number88403
Lot NumberP00091024 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number88403
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 3.5K MWCO > Cup, Conical; 2 mL; 25 Samples, 200–2000 μL each

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.