Learn More
Thermo Scientific™ Smart-Vue Pro Duo Modules 868Mhz
Increase efficiency with multi-sensor wireless monitoring
Brand:  Thermo Scientific™ SVPHWRMOD022
Product Code. 17591066
  • 694.00 € / Each
Explore more special offers
Description
Description
Smart-Vue Pro Duo 868 Mhz Only. Delivered with two 3.6 V lithium batteries, one sensor mounting kit, one mounting holder, one antenna and one 1.5 m/5 ft. flat cable.
  • Bright 2.4 in. color touch screen with multiple views
  • Alarms locally with real -time audio-visual alarms on the modules
  • Audio visual alarms for local notification
  • Communication locally via Bluetooth and to the system via LoRaLow
  • AC power consumption through USB port access
  • Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
  • Choice of wired and wireless sensors with automatic sensor detection
  • User traceability with customized levels of user access with encrypted password protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Module
3.6 V lithium battery
Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
LCD
2 Years
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Smart-Vue Pro Duo Modules 868Mhz >