missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Snap-on or Push-in Cap Glass Specimen Vials

Store and display specimens securely and easily. Fisherbrand™ Snap-on or Push-in Cap Glass Specimen Vials are available in a variety of sizes. Fisherbrand™ snap-on or screw-in polyethylene caps are sold separately.

Brand:  Fisherbrand™ 15384769

58.62 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 15384769

  • 70.80 € / Pack of 162
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

  • Manufactured from clear Type 1B neutral glass
  • Versatile capacity range from a variety of sizes
  • Can be used with either snap-on or push-in caps
  • Rolled rim for easy cap application
  • Particularly suitable for the storage of powders and dry samples
  • Fisherbrand™ Polyethylene Push-in or Snap-on Caps for Glass Specimen Vials sold separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Neutral Glass (Type 1B)
Snap On or Push In Cap
22 mm
23 mm
14 mL
48 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Snap-on or Push-in Cap Glass Specimen Vials > Capacity: 14mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.