missing translation for 'onlineSavingsMsg'
Learn More

Jaytec Glass™ Soda Glass Milk and Cream Pipettes

Soda Glass Milk and Cream Pipettes are for use with thick liquids.

Brand:  Jaytec Glass™ MP.681

 View more versions of this product

Product Code. 12303448

  • 5.25 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Blowout pipettes with wide bore jets for thick liquids
  • The last drop in the jet must be blown-out except where shown
  • MP.688 (BS.696) NOT to be blown out
Specifications

Specifications

Soda Lime Glass
Delivery of a precise volume in dairy testing
±0.02 mL
0.5, 1 mL
One-mark Blowout Pipette
Butyrometer
Non-sterile
2
Clear
355 mm
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Jaytec Glass™ Soda Glass Milk and Cream Pipettes > Soda Glass Milk and Cream Pipettes; Capacity: 0.5/1 mL; Tolerance: ±0.02 mL; Type: Two Mark Blowout Pipette

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.