Learn More
Jaytec Glass™ Soda Glass Milk and Cream Pipettes
Soda Glass Milk and Cream Pipettes are for use with thick liquids.
Brand:  Jaytec Glass™ MP.687
 View more versions of this product
Product Code. 12373448
  • 9.70 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blowout pipettes with wide bore jets for thick liquids
  • The last drop in the jet must be blown-out except where shown
  • MP.688 (BS.696) NOT to be blown out
Specifications
Specifications
Soda Lime Glass
Used to deliver a precise volume of Amyl Alcohol in dairy testing
±0.05 mL
1 mL
Amyl Alcohol Pipette
Butyrometer
Non-sterile
1
Clear
250 mm
Reusable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Jaytec Glass™ Soda Glass Milk and Cream Pipettes > Soda Glass Milk and Cream Pipettes; Capacity: 1 mL; Tolerance: ±0.05 mL; Type: Amyl Alcohol Pipette, BS.696