missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals

37.17 € - 549.00 €

Chemical Identifiers

CAS 7646-69-7, 8042-47-5
Molecular Formula HNa
Molecular Weight (g/mol) 24
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10297520
SDS View Documents
Thermo Scientific Acros
332141000
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10338750
SDS View Documents
Thermo Scientific Acros
332145000
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12307103
SDS View Documents
Thermo Scientific Acros
332140020
2 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.