Learn More
Labcold™ Sparkfree Freezer, 124L
The Labcold Sparkfree range has a specially manufactured sparkfree interior with all sources of potential ignition removed from the chamber. Anything that could cause a spark is located outside of the chamber making it an important part of laboratory safety.
Brand: Labcold™ RLVF0417
Description
- Sparkfree safety interior
- Manual defrost
- Door Lock
- Access Port
- Self closing door
- Temperature display
- High/Low temperature alarm
- Door Open alarm
- 3 shelves
Specifications
Underbench | |
-18°C to -25°C | |
3 |
124 L | |
Solid | |
600 x 600 x 830 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.