missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffer G (10X)

Ensure the optimum reaction conditions for restriction enzymes with Thermo Scientific™ 10X Buffer G, premixed with BSA for enhanced stability.

Brand:  Thermo Scientific™ BG5

15.88 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11899592

  • 22.05 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Our Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. This system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

Restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme, while AarI and BveI require an oligonucleotide (also supplied with the enzyme). Esp3I requires DTT.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10X
5 x 1.0 mL
Buffer G
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2807538 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberBG5
Lot Number2829654 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBG5
Lot Number2814702 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBG5
Lot Number2835075 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBG5
Lot Number2842119 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberBG5
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Buffer G (10X) > 5 x 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.