Learn More
Brady™ SPC™ Absorbents SitePoint Chemical Maintenance Kit
Ideal for moving spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required, which is especially ideal for maintenance activities, at construction sites and other temporary locations.
Brand:  Brady™ 309080
Product Code. 17811755
  • 1085.00 € / 1 set
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Loaded SitePoint Mobile Unit with suitable products for cleaning up general spills
  • Features two (200 mm) wheels for mobility and anchor points for fixed installation
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
  • Includes three anchor points on the base, allowing the unit to be bolted to the ground, to avoid trolley movement without permission
Specifications
Specifications
SPHS1-BB1-YE-CHEM
26.4 gal.
MDPE
Spill Kit
SitePoint Mobile Unit, Chemical Refill, Wall-mounted Roll Dispenser (50 cm Wide), Storage Unit for Gloves, Storage Unit for Goggles, A4 Sign Holder, Fixings
5.95 cm
75.5 cm
5.95 cm
16 kg
Yellow, White
100 L
Spill Kit
Beverage and Food Processing, Chemical, Healthcare, Maritime, Oil and Gas, Pharmaceutical, Warehouse
2.3 in.
29.724 in.
172.5 cm
35.3 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ SPC™ Absorbents SitePoint Chemical Maintenance Kit > SPC Absorbents SitePoint Chemical Maintenance Kit; Includes: SitePoint Mobile Unit, Chemical Refill, Wall-mounted Roll Dispenser (50 cm Wide), Storage Unit for Gloves, Storage Unit for Goggles, A4 Sign Holder, Fixings