Learn More
Description
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.
Solubility
Miscible with diethyl ether and acetone. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | °C to 75°C |
Density | 0.858 |
Boiling Point | 285°C (25 mm) |
Flash Point | 218°C (424°F) |
Assay Percent Range | 0.97 |
Refractive Index | 1.4955 |
Quantity | 100 g |
Sensitivity | Air and light sensitive |
Merck Index | 14,8768 |
Solubility Information | Miscible with diethyl ether and acetone. Immiscible with water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P10L031 | Certificate of Analysis | 12/03/2025 | B20944.22,B20944.36 |
Y25K025 | Certificate of Analysis | 04/12/2024 | B20944.22,B20944.36 |
Y27K030 | Certificate of Analysis | 02/12/2024 | B20944.22,B20944.36 |
Y04K044 | Certificate of Analysis | 14/11/2024 | B20944.22,B20944.36 |
10250573 | Certificate of Analysis | 30/08/2024 | B20944.22,B20944.36 |
Safety and Handling

- Squalene
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contains : Squalene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.