missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Funnel

Used for a wide range of applications. Bochem™ Stainless Steel Funnel has a conical shape.

Brand:  Bochem™ 8721

 View more versions of this product

Product Code. 10231641

  • 219.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Handle 
  • With air canal
  • Tube conical shape
  • 18/10 Stainless steel
Specifications

Specifications

150mm
30mm
Stainless Steel
155 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Stainless Steel Funnel > Height: 155mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.