Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Made from stainless steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has an 100 mm diameter.
50.95 € - 259.00 €
Specifications
Quantity | 1 Ea. |
---|---|
Type | Sieve |
Includes | Certificate of conformity and record card |
Diameter (Metric) Outer | 100 mm |
Frame Material | Stainless Steel |
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
11572143
|
Fisherbrand™
01/100SBW106um |
106 μm |
54.15 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
11582163
|
Fisherbrand™
01/100SBW1.25mm |
1.25 mm |
50.95 €
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
11592163
|
Fisherbrand™
01/100SBW1.40mm |
1.4 mm |
55.65 €
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
11592143
|
Fisherbrand™
01/100SBW150um |
150 μm |
55.69 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
11502153
|
Fisherbrand™
01/100SBW160um |
160 μm | N/A | N/A | N/A | |||||
11512153
|
Fisherbrand™
01/100SBW180um |
180 μm |
53.85 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
11522173
|
Fisherbrand™
01/100SBW2.00mm |
2 mm |
61.75 €
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
11522153
|
Fisherbrand™
01/100SBW200um |
200 μm |
54.20 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
11532153
|
Fisherbrand™
01/100SBW212um |
212 μm |
53.20 €
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
11542173
|
Fisherbrand™
01/100SBW2.50mm |
2.5 mm |
59.00 €
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
11582133
|
Fisherbrand™
01/100SBW25UM |
25 μm |
259.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
11552173
|
Fisherbrand™
01/100SBW2.80mm |
2.8 mm |
65.35 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11552153
|
Fisherbrand™
01/100SBW300um |
300 μm |
54.30 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
11562173
|
Fisherbrand™
01/100SBW3.35mm |
3.35 mm |
66.90 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
11592133
|
Fisherbrand™
01/100SBW38um |
38 μm |
85.10 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11582153
|
Fisherbrand™
01/100SBW400um |
400 μm |
54.75 €
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
11572153
|
Fisherbrand™
01/100SBW355um |
355 μm |
57.65 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
11592153
|
Fisherbrand™
01/100SBW425um |
425 μm |
56.25 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
11502163
|
Fisherbrand™
01/100SBW500um |
500 μm |
68.93 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
11512143
|
Fisherbrand™
01/100SBW53um |
53 μm |
68.70 €
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
11532143
|
Fisherbrand™
01/100SBW75um |
75 μm |
54.45 €
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
11532163
|
Fisherbrand™
01/100SBW710um |
710 μm |
57.80 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11512163
|
Fisherbrand™
01/100SBW600um |
600 μm |
57.80 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11522163
|
Fisherbrand™
01/100SBW630um |
630 μm |
57.80 €
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
11542163
|
Fisherbrand™
01/100SBW800um |
800 μm |
57.60 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
11542143
|
Fisherbrand™
01/100SBW80um |
80 μm |
54.75 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11552143
|
Fisherbrand™
01/100SBW90um |
90 μm |
53.20 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
11552163
|
Fisherbrand™
01/100SBW850um |
850 μm |
59.30 €
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
11562153
|
Fisherbrand™
01/100SBW315um |
315 μm |
51.70 €
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
11572163
|
Fisherbrand™
01/100SBW1.18mm |
1.18 mm |
60.80 €
Each |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
11502173
|
Fisherbrand™
01/100SBW1.60mm |
1.6 mm |
60.80 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11512173
|
Fisherbrand™
01/100SBW1.70mm |
1.7 mm |
57.80 €
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
11532173
|
Fisherbrand™
01/100SBW2.6mm |
2.36 mm |
55.05 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
11562143
|
Fisherbrand™
01/100SBW100um |
100 μm |
61.30 €
Each |
In Stock
Log in to see stock. |
||||||
11582143
|
Fisherbrand™
01/100SBW125um |
125 μm |
54.15 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11562163
|
Fisherbrand™
01/100SBW1mm |
1 mm |
68.36 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
11502143
|
Fisherbrand™
01/100SBW45um |
45 μm |
69.95 €
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
11522143
|
Fisherbrand™
01/100SBW63um |
63 μm |
61.65 €
Each |
In Stock
Log in to see stock. |
||||||
11542153
|
Fisherbrand™
01/100SBW250um |
250 μm |
54.45 €
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
Descripción
- 100 mm diameter test sieves
- Stainless steel frame
- Woven wire mesh
- Supplied with Certificate of Conformity and record card
Especificaciones
1 Ea. | |
Certificate of conformity and record card | |
Stainless Steel |
Sieve | |
100 mm | |
Woven Wire |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh