missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Test Sieves, 315 mm Dia., in Pore Size Greater than 10 mm
Brand: RETSCH 60.172.090000
This item is not returnable.
View return policy
Specifications
1 Each | |
Test Sieve | |
For screening aggregates, separation, fractioning, particle size determination | |
Stainless Steel |
90 mm | |
ISO 3310 and ASTM E 11, DIN 3310/2 | |
315 mm | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RETSCH Stainless-Steel Test Sieves, 315 mm Dia., in Pore Size Greater than 10 mm > Pore Size: 90 mm
Spot an opportunity for improvement?Share a Content Correction