Learn More
Fisherbrand™ Stainless Steel Crucible Tongs
With a flattened hinge.
Brand:  Fisherbrand™ 5701/2
 View more versions of this product
Product Code. 11835151
  • 17.10 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Bow pattern
  • Jaws have serrated edges
  • Available in a range of sizes
Specifications
Specifications
Bow pattern
Bent
Stainless Steel
1 Ea.
200 mm
Crucible Tongs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Crucible Tongs > Length: 200mm