Learn More
Thermo Scientific™ Sterilin™ Pipette Bulbs
Transfer liquids safely using glass pipettes and these natural rubber pipette bulbs.
Brand: Thermo Scientific™ TET0102
238.07 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Transfer liquids safely using glass pipettes and these natural rubber Sterilin™ Pipette Bulbs.
- Ideal for use with glass Pasteur pipettes
- Manufactured from natural rubber
- Volume drawn up: 0.22 cm3
Ordering Information
- Not available in all geographic regions
Specifications
Pipette bulbs; No. 10 Teat (Case of 100) | |
Glass Pasteur Pipette | |
Natural Rubber | |
Ideal for use with glass Pasteur pipettes |