Learn More
Cytiva Superose™ 6 Increase Columns
High-resolution size-exclusion chromatography for studying large proteins and complexes. Cytiva Lifescience™ Superose™ 6 Increase Columns delivers high quality in size exclusion chromatography (SEC).
Brand: Cytiva 29091598
Description
Superose™ 6 Increase pre-packed columns deliver the quality and high resolution in size exclusion chromatography (SEC) — even for large proteins and aggregates.
Increased performance
- Higher resolution than traditional Superose 6 columns
- Wide fractionation range (Mr from 5,000 to 5,000,000)
- Withstands high-pH cleaning

Specifications
Superose 6 Increase 3.2/300 | |
300 mm | |
Borosilicate glass | |
435 psi | |
8.6μm | |
4° to 30° in 20% ethanol |
3.2 mm | |
0.04mL/min. | |
Cross-linked agarose | |
3 to 12 | |
Titatium frit | |
Gel Filtration Column |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Superose 6 Increase 3.2/300
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.