Learn More
Sartorius Tacta™ Mechanical Pipettes
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
453.00 € - 1375.00 €
Specifications
Adjustable Tip Spacing | Yes |
---|---|
Autoclavable | Autoclavable |
Disposable | Reusable |
Grip Type | Ergonomic |
Material | Polypropylene |
Product Code | Brand | Number of Channels | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Number of Channels | Volume (Metric) | Color | Price | Quantity & Availability | ||||
15566304
![]() ![]() |
Sartorius
LH-729010 |
1 | 0.1 to 3 μL | Gray |
453.00 €
Each |
In Stock
Log in to see stock. |
||||
15576304
![]() ![]() |
Sartorius
LH-729020 |
1 | 0.5 to 10 μL | Gray |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15586304
![]() ![]() |
Sartorius
LH-729030 |
1 | 2 to 20 μL | Yellow |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15596304
![]() ![]() |
Sartorius
LH-729050 |
1 | 10 to 100 μL | Yellow |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15506314
![]() ![]() |
Sartorius
LH-729060 |
1 | 20 to 200 μL | Yellow |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15516314
![]() ![]() |
Sartorius
LH-729070 |
1 | 100 to 1000 μL | Blue |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15526314
![]() ![]() |
Sartorius
LH-729080 |
1 | 500 to 5000 μL | Green |
453.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
15536314
![]() ![]() |
Sartorius
LH-729090 |
1 | 1000 to 10,000 μL | Red |
453.00 €
Each |
In Stock
Log in to see stock. |
||||
15546314
![]() ![]() |
Sartorius
LH-729120 |
8 | 0.5 to 10 μL | Gray |
1215.00 €
Each |
In Stock
Log in to see stock. |
||||
15556314
![]() ![]() |
Sartorius
LH-729130 |
8 | 5 to 100 μL | Yellow |
1215.00 €
Each |
In Stock
Log in to see stock. |
||||
15566314
![]() ![]() |
Sartorius
LH-729140 |
8 | 30 to 300 μL | Orange |
1215.00 €
Each |
In Stock
Log in to see stock. |
||||
15576314
![]() ![]() |
Sartorius
LH-729220 |
12 | 0.5 to 10 μL | Gray |
1375.00 €
Each |
In Stock
Log in to see stock. |
||||
15586314
![]() ![]() |
Sartorius
LH-729230 |
12 | 5 to 100 μL | Yellow |
1375.00 €
Each |
In Stock
Log in to see stock. |
||||
15596314
![]() ![]() |
Sartorius
LH-729240 |
12 | 30 to 300 μL | Orange |
1375.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Comfortable to hold due to the ergonomically designed handle
- Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
- Controlled and smooth tip ejection with the new Sartorius Optiject technology
- Calibration adjustment to provide accurate results for various liquid types
- Easy to clean, with only three parts to disassemble
- Reliable results, even over lengthy pipetting periods
Specifications
Yes | |
Reusable | |
Polypropylene | |
No | |
Pipette |
Autoclavable | |
Ergonomic | |
Tacta | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Tacta™ Mechanical Pipettes