missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ qPCR Master Mix, CG
GREENER_CHOICE

TaqPath qPCR Master Mix, CG, is a 2X formulation designed for gene expression and miRNA analysis. It contains thermostable Fast DNA polymerase, uracil-N-glycosylase (UNG), dNTPs with dUTP, ROX dye (passive reference), and optimized buffer components for maximum robustness and reproducibility.

Brand:  Applied Biosystems™ A16245

755.70 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16258100

  • 1374.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store at 2°C to 8°C in the dark.
Liquid
qPCR
Fast or Standard
ROX (Pre-mixed)
qPCR Master Mix
1000 Reactions
2X
1000 Reactions
2 x 5 mL
Primer-probe
Bottle
Fast DNA Polymerase
QuantStudio™ Dx, 7500 Fast Dx
TaqPath™
2 x 5 mL
Gene Expression, Virus Detection
Built-In Hot Start
cDNA, DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqPath™ qPCR Master Mix, CG > 2 x 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.