missing translation for 'onlineSavingsMsg'
Learn More
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals Product Code.: 15678725

Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals

Product Code. 15678725
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15678725

Brand: Thermo Scientific Acros 461250050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202138-50-9
Molecular Formula C19H30N5O10P·C4H4O4
Molecular Weight (g/mol) 635.51
InChI Key VCMJCVGFSROFHV-WZGZYPNHSA-N
Synonym tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1Show MoreShow Less
PubChem CID 6398764
ChEBI CHEBI:63718
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acidShow MoreShow Less
SMILES CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: >10 mg/mL.
Formula Weight 635.51
Percent Purity 98%
Chemical Name or Material Tenofovir disoproxil fumarate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451984 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number461250250,461250050,461250010
Lot NumberA0374687 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number461250250,461250050,461250010
Lot NumberA0448232 Certificate TypeCertificate of Analysis Date09/12/2022 Catalog Number461250250,461250050,461250010
Lot NumberA0445553 Certificate TypeCertificate of Analysis Date02/09/2022 Catalog Number461250250,461250050,461250010
Lot NumberA0444631 Certificate TypeCertificate of Analysis Date03/08/2022 Catalog Number461250250,461250050,461250010
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.