missing translation for 'onlineSavingsMsg'
Learn More
Tetrabutylammonium iodide, 98%, Thermo Scientific Chemicals Product Code.: 11985441

Tetrabutylammonium iodide, 98%, Thermo Scientific Chemicals

Product Code. 11985441
2.5 kg, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 11985441

Brand: Thermo Scientific Acros 161270025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 311-28-4
Molecular Formula C16H36IN
Molecular Weight (g/mol) 369.38
MDL Number MFCD00011636
InChI Key DPKBAXPHAYBPRL-UHFFFAOYSA-M
Synonym tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodideShow MoreShow Less
PubChem CID 67553
IUPAC Name tetrabutylazanium;iodide
SMILES [I-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 141.0°C to 150.0°C
Color White to Cream
Ignition Residue 1% max.
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula [CH3(CH2)3]4NI
Quantity 2.5 kg
Fieser 05,646; 06,566; 07,355; 09,447; 11,501
Solubility Information Solubility in water: <5%.
Formula Weight 369.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466068 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number161275000,161270250,161271000,161270025
Lot NumberA0458917 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number161275000,161270250,161271000,161270025
Lot NumberA0452889 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number161275000,161270250,161271000,161270025
Lot NumberA0450592 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number161275000,161270250,161271000,161270025
Lot NumberA0439805 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number161275000,161270250,161271000,161270025
compliance-icons
Product Identifier
  • Tetrabutylammonium iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabutylammonium iodide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.