Learn More
Texwipe™ AlphaMop™
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
Brand: Texwipe™ TX7108
Description
Specifically designed for the needs of ultraclean production environments. Easily changeable, laundered polyester mop covers fit over a flat, foam-padded head assembly. Lightweight, durable fiberglass handle provides excellent control over swivel head and accesses hard-to-reach areas with ease.
- Enables rapid change of wiping surfaces for adherence to strict wiping protocol ISO Class 4 cleanroom-laundered Alpha™ polyester mop cover
- Replaceable foam pad ensures that the polyester mop cover conforms to textured floors and other surfaces
- Maintains ultralow generation of particles and extractables Resists corrosion, abrasion and commonly used cleaning and disinfecting chemicals such as bleaches, phenols and quaternary ammonium compound
- AlphaMop polyester replacement mopheads and foam pads available separately (150 heads and six pads per case)
- AlphaMop mop covers presaturated with 6% IPA Honeycomb™ 10 double-knit polyester mop covers have an abrasive texture (supplied 150 covers per case)
Specifications
Cleanroom Mop | |
Autoclavable | |
Ideal for applications requiring low particle, extractable and ionic contamination, superior for wall, floor and ceiling cleaning, designed specifically for ultraclean manufacturing environments |
Non-sterile | |
60 in. Fiberglass Handle, 15 x 8 in. Single Head Assembly, 6 Polyester Alphawipe Mop Covers | |
Polyester |