missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theophylline, 99+%, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 270.0°C to 274.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15, 9436 |
Solubility Information | Solubility in water: 8.3g/L (20°C). Other solubilities: soluble in alkaline solutions, slightly soluble in ethanol and chloroform |
Formula Weight | 180.17 |
Percent Purity | 99+% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0453328 | Certificate of Analysis | 25/01/2025 | 250310050,250312500,250310010 |
A0453328 | Certificate of Analysis | 25/08/2023 | 250310050,250312500,250310010 |
A0427710 | Certificate of Analysis | 05/09/2022 | 250310050,250312500,250310010 |
A0404191 | Certificate of Analysis | 16/07/2020 | 250310050,250312500,250310010 |
Safety and Handling


Product Identifier
- Theophylline
- Danger
- Acute toxicity Category 3
- Reproductive toxicity Category 1B
- H301-Toxic if swallowed.
- H360D-May damage the unborn child.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Theophylline
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Theophylline, 99+%, anhydrous, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction