Learn More
Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
Brand: Thermo Scientific™ 2130
442.26 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
Stainless Steel:- Durable, corrosion-resistant stainless-steel exterior
- Carrying handle and vented secure lid
Shallow Wide Mouth:
- Ideal for use as a cold trap
- Convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol
- Stainless-steel jacket and cushioned base for years of trouble-free use
Plastic Wide Mouth:
- Low cost, high efficiency
- Rugged, shock-absorbing plastic exterior with carry handle
- “Push-fit” insulated lid, vented to prevent pressure build-up
Enameled Steel:
- Safe storage of liquefied gases
- Cushioned base
- Models 2118-2121 have insulated lids and fold-down carrying handles
- Ideal for use as a cold trap

Specifications
1.9 L | |
Benchtop | |
135 mm | |
155 mm |
Stainless Steel | |
Without Lid and Handle | |
186 mm | |
4.5 kg |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.