Learn More
Thermo Scientific™ Disposable Inoculator Assembly for 96-Well Plates
GREENER_CHOICE
For bacterial studies, blood toxicity and many tissue culture applications
Brand:  Thermo Scientific™ 12-0050
380.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10685757
  • 396.00 € / Pack of 50
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For bacterial studies, blood toxicity and many tissue culture applications
Simultaneously transfers bacterial colonies from a vessel of choice to a 96-well Microtiter™ format.
  • Sterilized by gamma radiation
  • For use with inoculum tray
  • Includes 95 tips and one control/blank
  • 10μL/tip
Designed for bacterial studies, blood toxicity and many tissue culture applications.
Order Info
Case of 50
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Disposable Inoculator Assembly for 96-Well Plates > Disposable Inoculator Assembly