Learn More
Thermo Scientific™ Remel™ Bacillus cereus ATCC™ 11778™, Culti-Loops™
Thermo Scientific Remel Bacillus cereus ATCC 11778, Culti-Loops, ATCC: 11778TM, Species: Bacillus cereus, Recommended Applications: Healthcare, Strain Designations: FDA strain PCI 213 [ATCC 9634, NCTC 10320, S.A. Waksman strain O]
Brand: Thermo Scientific™ R4601220
140.59 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Thermo Scientific Remel Bacillus cereus ATCC 11778, Culti-Loops, ATCC: 11778TM, Species: Bacillus cereus, Recommended Applications: Healthcare, Strain Designations: FDA strain PCI 213 [ATCC 9634, NCTC 10320, S.A. Waksman strain O]
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Certifications
Culti-Loops are CE marked.

Specifications
Bacillus cereus | |
Culti-Loop | |
FDA strain PCI 213 [ATCC 9634, NCTC 10320, S.A. Waksman strain O] |
11778™* | |
5 Loops/Pack |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
193213 | Certificate of Analysis | 18/12/2024 | R4601220 |
162584 | Certificate of Analysis | 12/09/2024 | R4601220 |
141341 | Certificate of Analysis | 25/07/2024 | R4601220 |
106538 | Certificate of Analysis | 09/05/2024 | R4601220 |
842248 | Certificate of Analysis | 16/04/2024 | R4601220 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.