missing translation for 'onlineSavingsMsg'
Learn More
Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals Product Code.: 11429290

Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals

Product Code. 11429290
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11429290

Brand: Thermo Scientific Alfa Aesar H50172.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107818-20-2
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD01863636
InChI Key AACVULYSNJAKEQ-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8pShow MoreShow Less
PubChem CID 5074765
IUPAC Name 4H-thieno[3,2-b]pyridin-7-one
SMILES C1=CNC2=C(C1=O)SC=C2

Specifications

Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 151.19
Percent Purity 96%
Chemical Name or Material Thieno[3,2-b]pyridin-7-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD22Z022 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberH50172.03,H50172.06
Lot NumberH26W009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50172.03,H50172.06
Lot NumberH09X048 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50172.03,H50172.06
Lot NumberH19U027 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50172.03,H50172.06
Lot NumberD22Z022 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50172.03,H50172.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.