missing translation for 'onlineSavingsMsg'
Learn More

Total Organic Carbon (TOC) Standard 500 ppm
BIOPHARMA

Manufactured from high purity raw materials and suitable for calibrating a wide range of TOC analyzers to a high accuracy

Brand:  Reagecon TOC500

Product Code. 11556633

  • 119.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Total Organic Carbon (TOC) Reference Standard, Total Organic Carbon (TOC) Reference Material.
  • Independent, traceable and certified.
  • Presented in a specially cleaned and leached amber glass bottle.
  • Prepared and certified gravimetrically on a weight/volume basis to a specification of ± 2%.
  • Reagecon holds IEC/ISO 17025 accreditation for calibration of laboratory balances and pipettes (INAB Ref: 265C).
  • The resulting equipment calibration certificates are issued in accordance with the requirements of ISO/IEC 17025.
  • The concentration of this standard is verified using a high performance calibrated total organic carbon analyzer (TOC Instrument).
  • The calibration of the TOC instrument is completed using a high purity ISO Guide 34 accredited TOC standard similar in TOC concentration value to this product.
Specifications

Specifications

Accuracy ± 2&, Matrix: Water (H2O)
500 mL
TOC Analysis
HDPE Twin Neck Bottle
500 ppm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberTOC50025B1 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberTOC500
Lot NumberTOC50024K1 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberTOC500
Lot NumberTOC50024F1 Certificate TypeCertificate of Analysis Date08/10/2024 Catalog NumberTOC500
Lot NumberTOC50024D1 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog NumberTOC500
Lot NumberTOC50024B1 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog NumberTOC500
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Total Organic Carbon (TOC) Standard 500 ppm > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.