missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Translucent Silicone Tubing

Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.

Brand:  Fisherbrand™ 1187-6293

 View more versions of this product

Product Code. 11876293

  • 110.00 € / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Translucent silicone tubing
  • FDA compliant (food)
Specifications

Specifications

FDA, BFR and WRAS approved
4 mm
10 mm
3 mm
Pharmaceutical , biotechnology, transfer sterile fluids
Sterile
Flexible
Autoclavable
Silicone
5 m
Peristaltic pumps
-40°C
Translucent
250°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Translucent Silicone Tubing > I.D.: 4mm; O.D.: 10mm; Length: 5m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.