Learn More
Manutan Trigger operated water gun, 150/10 psi/bar, 3/4 inch BSP (F)
Brand:  Manutan A327222
Encompass_Preferred
Product Code. 12813889
Description
Description
Fingertip control of water usage in a variety of wash down applications. Adjustable spray/jet pattern with automatic water shut-off on release of trigger.
  • Working pressure : Max of 150psi
  • Working pressure : Max of 10bar
  • Max. flow rate : approx. 15L/min
  • 3/4 BSP (F) fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan Trigger operated water gun, 150/10 psi/bar, 3/4 inch BSP (F)