missing translation for 'onlineSavingsMsg'
Learn More

TPP Techno Plastic Products AG TubeSpin™ Bioreactor

TubeSpin™ Bioreactor, suspension cells

Brand:  TPP Techno Plastic Products AG 80600

 View more versions of this product

Product Code. 17251766

  • 105640.00 € / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • TPP TubeSpin™ Bioreactors have proven their advantages in large scale screening and optimizing of suspension cell system process development.
  • The important parameters for the production upscaling can be evaluated with small amounts maximizing the number of parallel experiments at the same time. This lowers the costs and increases the efficiency of the process
  • TubeSpin™ Bioreactor 15: working volume 1 - 15 mL, conical or round bottom, filter cap
  • TubeSpin™ Bioreactor 50: working volume 1 - 35 mL, conical or round bottom, filter cap
  • TubeSpin™ Bioreactor 450: working volume up to approx. 300 ml, conical bottom, filter cap, additional replacement cap available: solid #87355, filter #87356
  • TubeSpin™ Bioreactor 600: working volume up to approx. 400 ml, conical bottom, filter cap, additional replacement cap available: solid #87355, filter #87356
  • Filter cap with partly 5 different openings (A, B, C, D, E) or 10 equal openings
  • Fit standard centrifuge apparatus
  • Matching adapter plates for shakers for cultivation of suspension cells available
Specifications

Specifications

600 mL
Polypropylene
Single-use
Conical, Filter screw cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TPP Techno Plastic Products AG TubeSpin™ Bioreactor > Bioreactor; 600 mL; conical; filter screw cap; 3500 x g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.