missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TurboFect™ Transfection Reagent

Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.

Brand:  Thermo Scientific™ R0533

64.46 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 12395613

  • 73.25 € / 5 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.

TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery into eukaryotic cells. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells. Transfection can be performed in the presence or absence of serum. TurboFect Reagent demonstrates superior transfection efficiency and minimal toxicity when compared to other lipid-based or polymer-based transfection reagents.

Highlights:

  • High transfection efficiency of a wide variety of cell types
  • No optimization required for most cell lines
  • Excellent transfection efficiency in the presence or absence of serum
  • Minimal cytotoxicity
  • Ready-to-use – no need to reconstitute, dilute or manipulate

Recommended for:

Stable and transient transfection or co-transfection of DNA or RNA; Experiments with adherent or suspension cells, differentiated or undifferentiated cells; Gene expression studies and assays involving GFP or luciferase reporters

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transfection Reagent
200 μL
Lipid-Based Transfection
100 Transfections
Upon receipt store at 4°C.
TurboFect™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2851444 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberR0533
Lot Number2818562 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0533
Lot Number2869981 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0533
Lot Number2923854 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberR0533
Lot Number2961908 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberR0533
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TurboFect™ Transfection Reagent > 200 μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.