missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem 2000 C

Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).

Brand:  DuPont™ D13395589

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11733093

  • 574.00 € / Pack of 25
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hooded coverall with elastic face opening
  • Adhesive chin flap for tight fit around respirator
  • Stitched and over-taped seams
  • Souble self-adhesive zipper flap
  • Elastic cuffs, ankles and waist
  • Thumb loops prevent sleeves from riding up
  • 100% particle tight fabric
  • Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
  • Antistatic treatment (inside)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
Tychem
Elastic
Thumb Loop
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
EN 14126, EN 1073-2, EN 1149-5
Used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
Large
Yellow
Elastic
Elastic
Stitched, Taped
Polymeric barrier coating
100 to 108 cm
174 to 182 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tychem 2000 C > Size: Large

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.