Learn More
DuPont™ Tychem 2000 C
Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
24.15 € - 574.00 €
Specifications
Product Type Hooded Coveralls
Material Tychem
Color Yellow
Waist Style Elastic
Ankle Style Elastic
View More Specs
Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12658341
View accessory products View Documents
DuPont™
D13494969
Medium
574.00 €
Pack of 25
In Stock
to see stock.
Add to basket
12369419
View accessory products View Documents
DuPont™
D13494969
Medium
24.15 €
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
12379419
View accessory products View Documents
DuPont™
D13494990
Small
24.15 €
Each
In Stock
to see stock.
Add to basket
12648341
View accessory products View Documents
DuPont™
D13494990
Small
574.00 €
Pack of 25
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
11743093
View accessory products View Documents
DuPont™
D13395699
X-Large
574.00 €
Pack of 25
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
12389419
View accessory products View Documents
DuPont™
D13395699
X-Large
24.15 €
Each
In Stock
to see stock.
Add to basket
12359419
View accessory products View Documents
DuPont™
D13395589
Large
24.15 €
Each
In Stock
to see stock.
Add to basket
11733093
View accessory products View Documents
DuPont™
D13395589
Large
574.00 €
Pack of 25
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
12932159
View accessory products View Documents
DuPont™
D13395560
2X-Large
27.15 €
Each
In Stock
to see stock.
Add to basket
11753093
View accessory products View Documents
DuPont™
D13395560
2X-Large
574.00 €
Pack of 25
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • Hooded coverall with elastic face opening
  • Adhesive chin flap for tight fit around respirator
  • Stitched and over-taped seams
  • Souble self-adhesive zipper flap
  • Elastic cuffs, ankles and waist
  • Thumb loops prevent sleeves from riding up
  • 100% particle tight fabric
  • Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
  • Antistatic treatment (inside)
Specifications
Specifications
Hooded Coveralls
Yellow
Elastic
Elastic
Stitched, Taped
EN 14126, EN 1073-2, EN 1149-5
Tychem
Elastic
Thumb Loop
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DuPont™ Tychem 2000 C