Learn More
Biochrom™ Ultrospec 7500 UV-Vis Spectrophotometer
The Ultrospec 7500 is a simple-to-use instrument with highly advanced performance functionality and a sophisticated GLP mode to ensure high quality, consistent and reliable results. Versatile and precise, this instrument is ideal for use in any manufacturing or production laboratory.
Brand:  Biochrom™ 80-2140-60
9376.50 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Product Code. 17717359
  • 9870.00 € / Each
Explore more special offers
Description
Description
  • Easy to use icon driven touch screen for true 'plug and play' setup
  • Automated 8-cell sample carousel supplied as standard
  • Robust, compact and reliable for general or heavily regulated laboratory use
  • Combines basic measurement modes with enhances software functionality and method development capabilities
  • Can be upgraded with Resolution CFR PC control software to comply with 21 CFR Part 11
  • Provides stored methods for DNA, RNA, oligo and protein quantification, as well as checking sample purity
  • Preloaded methods for commonly available fluorescent dyes for previous samples
  • Screenshot function to easily capture and export results
  • 800 x 480 pixel backlit, color LCD touch display
  • Xenon flash lamp for longer lifetime and lower running costs
  • IQ/OQ available on request
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Biochrom™ Ultrospec 7500 UV-Vis Spectrophotometer > Ultrospec 7500 UV-Vis Spectrophotometer; Dimensions (L x W x H): 510 x 350 x 160 mm; Optical Design: Split Beam Optics; Product Line: Ultrospec 7000; Spectral Bandwidth: <2 nm; Wavelength Range: 190 to 1100 nm