Learn More
BICASA Undercounter Drawer Cabinet on Casters
Undercounter Drawer Cabinet on Casters with fire-retardant particleboard body and drawers with soft closing and single lock.
Brand:  BICASA TN.DW.0600.630-B
 View more versions of this product
Product Code. 30057581
  • 691.00 € / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Undercounter cabinets on casters
  • Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads), 2 with lockable with brakes - Height mm 100
  • Central locking system as standard
  • Anti-tilt safety system to open only one drawer at a time
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
Specifications
Specifications
Mobile Storage Cabinet
Underbench cabinet
1
23.6 in.
Yes
Blue
600 x (500 + 20) x 630 mm
3
600 mm
Melamine
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BICASA Undercounter Drawer Cabinet on Casters > Dimensions (L x W x H): 600 x (500 + 20) x 630 mm, Color Facade: Blue