missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Upper Basket for Narrow Necked Glassware

Upper basket O 187 for the optimum loading of narrow necked glassware

Brand:  Miele Professional™ 3881640

2829.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10699892

  • 2990.00 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Equipped with 34 injector nozzles
  • Vertical clearance 170 mm
  • Powder dispensing not possible
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Upper Basket/Injector Unit
475 mm
531 mm
O 187
250 mm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Miele Professional™ Upper Basket for Narrow Necked Glassware > Dimensions (D x W x H): 475 x 531 x 250mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.