Learn More
Fisherbrand™ Stainless Steel Freezer Rack

Store assay plates and PCR racks securely in steel freezer racks. Stainless Steel Freezer Racks are available in two styles: locking rod racks that allow stacking of assay plates or horizontal drawer racks for easy box access. Both styles provide 16 compartments of secure cryostorage.
Brand: Fisherbrand™ HS2864
Description
For cryostorage in upright freezers.
- Constructed of corrosion-resistant stainless steel.
- For Assay Plate, PCR rack and cryogenic tube box storage.
- Designed to fit in most standard upright freezers.
- Sturdy containers with 4 levels of storage, providing 16 total storage compartments.
- Wire handles on the front allow for easy removal of the rack from the freezer.
Locking Rod Freezer Rack:
- Rack includes four vertical locking rods over the side opening to secure contents in place.
- Allows stacking of short assay plates in each slot (not included):
- 96-Well Assay plates (holds 3 per compartment, up to 48 total)
- 24-Well Assay plates (holds 2 per compartment, up to 32 total)
- 96-Deep Well Assay plates (holds 1 per compartment, up to 16 total)
- 96-Well PCR racks (holds 1 per compartment, up to 16 total)
Horizontal Drawer Freezer Rack
- Horizontal drawer system allow for easy box access
- Drawers feature a stop that pivots down when fully extended
- Polycarbonate drawer handles provide a user-friendly grip
- Includes a slot and label for drawer/sample identification
- Ideal for standard boxes up to 133 x 133 x 53 mm in size
Specifications
Freezer Rack | |
Not Autoclavable | |
Boxes (2 in.) | |
16 | |
Pivoting Pull-out Drawers | |
Non-sterile | |
4 | |
1 Ea. | |
571 mm | |
139 mm | |
238 mm |
4 x 4 | |
Silver | |
Stainless Steel | |
Rectangular | |
Yes | |
For standard PCR rack and cryogenic rack (up to 133 x 133 x 53 mm) storage in upright freezers | |
1 Each | |
571 x 139 x 238 mm | |
22.5 in. | |
5.5 in. | |
9.4 in. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.