missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Water purification

Stand fitting E 314 for the manual production of demineralised water

Brand:  Miele Professional™ 3830340

436.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10732223

  • 494.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pressurised to 10 bar
  • Complete with pressure hose 150 cm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
Stand fitting
Plastic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Miele Professional™ Water purification > Stand fitting

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.