Learn More
Thermo Scientific™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments
Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments are for use with ACC- and WPS-3000 autosamplers.
Brand:  Thermo Scientific™ 6820.4079
 View more versions of this product
Product Code. 13472018
  • 509.00 € / Each
Explore more special offers
Description
Description
Use Thermo Scientific™ Dionex™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments with ACC- and WPS-3000 autosamplers.
Specifications
Specifications
Low Profile Support Rack
Support rack for deep-well plate, 34-46 mm height for all WPS-3000 variants
Deep-Well Plates for All WPS-3000 LC Autosamplers (Not ACC)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments >