missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WypAll™ L10 Extra Wiper Centrefeed Roll 7140, 1 Roll x 1,500 Paper Wipers
GREENER_CHOICE

L10 is deal for light duty wiping and perfect for tasks such as polishing glass, cleaning surfaces, and tools. The single use wipes with Airflex™ technology deliver exceptional strength and absorbency, even when wet, and help break the chain of germ transmission.

Brand:  Kimberly-Clark™ 7140

31.43 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 12680533

  • 55.10 € / 1 roll
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • L10 wiping rolls contain 1 ply wiping paper sheets
  • Dry cleaning wipes are perfect for light duty wiping, such as polishing glass, cleaning surfaces and tools
  • Airflex base sheet technology of these paper wipes is strong enough to see any job through from start to finish without falling apart and absorbent enough to clean up most spills with ease
  • Large centrefeed wiper roll format ideal for larger manufacturing and industrial work areas, or anywhere requiring extensive wiping
  • Wipers are compatible with a range of Kimberly-Clark Professional rolled hand towel dispensers, including product code 7017
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wiping Paper
30 cm
30 cm
Blue
Everyday cleaning, frequent light wiping and small spills, being stronger than conventional 1-ply wipers
Non-sterile
11.81 in.
11.81 in
Paper
38 x 23.5 cm
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ WypAll™ L10 Extra Wiper Centrefeed Roll 7140, 1 Roll x 1,500 Paper Wipers > Color: Blue, Dimensions (L x W): 38 x 23.5 cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.