missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™ Product Code.: 10357684

1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10357684
25 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10357684

Brand: Thermo Scientific Maybridge SPB00444EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3290-06-0
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
InChI Key ZFLRKAMKGYNFPH-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl chloride, 1,3-dichloro-5-chloromethyl benzene, benzene, 1,3-dichloro-5-chloromethyl, 3,5-dichlorobenzylchloride, unii-c076i19eql, 5-chloromethyl-1,3-dichlorobenzene, pubchem16695, alpha,3,5-trichlorotoluene, 3,4-dichloro benzyl chloride, ksc495g8lShow MoreShow Less
PubChem CID 137880
IUPAC Name 1,3-dichloro-5-(chloromethyl)benzene
SMILES C1=C(C=C(C=C1Cl)Cl)CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2Cl
Quantity 25 g
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-5-(chloromethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.